Pesticide reference standard, Chlorpyrifos-methyl 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERXA11601000CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:5598-13-0
- Description:Chlorpyrifos-methyl 100 µg/ml in cyclohexane
- Formula:C₇H₇Cl₃NO₃PS
- Description:Chlorpyrifos-methyl