Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
4-Nitrophenyl phosphate

4-Nitrophenyl phosphate

100 mL
Catalog # ENZOADI800076
Corrosive
Irritant
Health
Supplier:   Enzo Life Sciences
4-Nitrophenyl phosphate
Catalog # ENZOADI800076
Supplier:  Enzo Life Sciences
CAS Number:  330-13-2
4-Nitrophenyl phosphate

100 mL

CAS Number:  330-13-2
Price
£536.00
Log in for availability
1 * 100 mL
Product Details & Documents

AP substrate.

  • Used in ELISA, colorimetric detection
  • Solution of p-nitrophenylphosphate in buffer
Synonyms:  4-Nitrophenyl dihydrogen phosphate, pNpp, p-Nitrophenyl phosphate
  • CAS Number:330-13-2
  • Formula:C₆H₆NO₆P
  • Molecular Weight:219.09 g/mol
  • Storage Temperature:Freezer
Specifications

Specifications

  • Pk:
    100 mL

Specifications