Aflatoxin B1 ≥98% (by HPLC, TLC)
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
CAS Number:1162-65-8
:
Formula:C₁₇H₁₂O₆
:312.3 g/mol
Merck Index:14,00180
Storage Temperature:Refrigerator
EINECS:214-603-3
UN:2811
ADR:6.1,I
:GHS06
:GHS08
Specification Test Results
- Source:Isolated from Aspergillus flavus
- Purity:≥98% (HPLC, TLC)
Specification Test Results
Frequently Bought Together