Order Entry
United Kingdom
ContactUsLinkComponent
 
Flunarizine dihydrochloride ≥99%
Catalog # ENZOALX550268G001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Flunarizine dihydrochloride ≥99%
Catalog # ENZOALX550268G001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :477.40 g/mol
CAS Number:30484-77-6
Formula:C₂₆H₂₆F₂N₂·2HCl
MDL Number:MFCD00058198