Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
RWJ-60475 (AM)3
RWJ-60475 (AM)3
RWJ-60475 (AM)3
RWJ-60475 (AM)3
Catalog # ENZOBMLPR1100001
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Expand image
RWJ-60475 (AM)3
Catalog # ENZOBMLPR1100001
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Specifications
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
:
620.4 g/mol
Formula:
C₂₂H₂₃BrNO₁₃P
Product Details