Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ozagrel
Ozagrel ≥98%
Ozagrel ≥98%
Ozagrel ≥98%
Catalog # ENZOBMLEI3780010
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Expand image
Ozagrel ≥98%
Catalog # ENZOBMLEI3780010
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Specifications
Pk:
10 mg
Seq:
2275
Specifications
About this item
Product Details
:
:
228.2466 g/mol
CAS Number:
82571-53-7
Formula:
C₁₃H₁₂N₂O₂
MDL Number:
MFCD00868231
Product Details