Microplate washer, Wellwash™
Wash 96-well plates securely with the Thermo Scientific™ Wellwash™ Microplate Washer, an easy-to-use, reliable microplate-strip washer for routine ELISA applications. The Wellwash features a graphical user interface, local language versions and USB port for optimal ease-of-use and convenience.
- Large color clear display and keypad for easy, convenient use
- User interface is logic and intuitive for quick protocols development
- Easy-to-use software requires minimal training for fast startup
- Unpressurized bottles are safe and secure to use
- Liquid level sensors are present in both wash and waste bottle for security
- Automatic prime feature assure safe performance
- Aerosol cover prevents aerosolization of infectous diseases
- Plate sensor recognizes if plate is present
- Wellwash is designed to efficiently remove the excess liquid from the wells
- Low residual volumes provide optimal washing performance and reliable assay results
Recommended for ELISA-based assays.
: One-year warranty.
: Includes 1×2 L wash bottle, 1×2 L waste bottle, 1×8 or 1×12 wash head, aerosol cover.
- Catalog No:
- 736-0422
- 736-3008
- Model:
- Wellwash
- Wellwash
- Plate types:
- 96-well
- 96-well
- Wash heads:
- 1×8
- 1×12
- Washing volume:
- 50 - 1000 μl
- 50 - 1000 μl
- Dispensing volume range:
- 50 - 400 µl
- 50 - 400 µl
- Display:
- Colour LCD
- Colour LCD
- Interfaces:
- USB
- USB
- Priming and rinsing volume:
- 5 - 100 ml
- 5 - 100 ml
- Residual aspiration volume:
- <2 μl
- <2 μl
- W×D×H:
- 345×385×240 mm
- 345×385×240 mm
- Weight:
- 8 kg
- 8 kg
Frequently Bought Together