

[4-Chloro-6-(2,3-xylidino)-2-pyrimidinylthio]acetic acid ≥97% (by HPLC)
ENZOBMLGR2000250
:
- Pk:250 mg
- Seq:2590
- Purity:≥97% (HPLC)
- Identity:Determined by NMR
- Appearance:White to off-white solid
:
:323.80 g/mol
:155…157 °C
:GHS07
:GHS08
:Ambient
:50892-23-4
:C₁₄H₁₄ClN₃O₂S
:MFCD00191335