Order Entry
United Kingdom
ContactUsLinkComponent
Nitric acid 0.2 mol/l (0.2 N)
Irritant
Nitric acid 0.2 mol/l (0.2 N)
Catalog # VWRS5550.5000
Supplier:  VWR Collection
CAS Number:  
Irritant
Nitric acid 0.2 mol/l (0.2 N)
Catalog # VWRS5550.5000
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pack type:
    Plastic container
  • Pk:
    5 L
  • Seq:
    5190

Specifications

About this item

 :
 :63.01 g/mol
Storage Temperature:Ambient
EINECS:231-714-2
UN:2031
ADR:8,II
 :GHS07
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349