Barium hydroxide 0.15 mol/l (0.3 N) in aqueous solution, Sigma-Aldrich®
Catalog # SIALB4059-500ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
About this item
:
:171.34 g/mol
:M-Clarity™ quality level = MQ200
EINECS:241-234-5
CAS Number:17194-00-2
Formula:H₂BaO₂
MDL Number:MFCD00003443