4-Pyridineboronic acid
Glass bottle for solids, 5 g
Catalog # FLUO010820-5G
Marketsource
Supplier: FLUOROCHEM
4-Pyridineboronic acid
Catalog # FLUO010820-5G
Supplier: FLUOROCHEM
Marketsource
CAS Number: 1692-15-5
Jump to:
Product Details & Documents
Synonyms: 4-Pyridinylboronic acid, Pyridine-4-boronic acid, Pyridin-4-ylboronic acid
- CAS Number:1692-15-5
- Formula:C₅H₆BNO₂
- MDL Number:MFCD01074545
- Molecular Weight:122.92 g/mol (anhy)
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



