0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Electrophoresis Systems
Horizontal Gel Electrophoresis Systems
46320561
undefined | Accessories
CLEVGCI-HZ58-CST
:
CLEAVER SCIENTIFIC
CLEVGCI-HZ58-CST
:
CLEAVER SCIENTIFIC
Description:
Stand alone molded casting tray, removable gel tray with 3 visualisation strips, 2 gel casting dams
L×W×H:
204×100×25 mm
Pk:
1
For:
Horizon H58