Order Entry
United Kingdom
ContactUsLinkComponent
o-Tolidine ≥98%
o-Tolidine ≥98%
  A17837.14
 :  
o-Tolidine ≥98%
  A17837.14
 :  
  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
 :
 :212.29 g/mol
 :14,9514
 :300…301 °C
 :128…132 °C
 :∼1.234 g/cm³ (20 °C)
 :244 °C (471 °F)
 :204-358-0
 :3077
 :9,III
 :GHS08
 :GHS07
 :GHS09
 :Ambient
 :119-93-7
 :[-C₆H₃(CH₃)-₄-NH₂]₂
 :MFCD00014773