ProSpore® Ampoule Biological Indicator
MESL77-02-PS-5-50
- Description:Incubation: 48 hours at 55...60 °C, population: 10⁵
- Environmentally Preferable:
- Pk:50
ProSpore® ampoule is a self-contained biological indicator available for use in steam sterilisation, and appropriate for use in healthcare applications.
- It is intended for use as a challenge to steam sterilization at 121 °C
- Ideal for use in validating the sterilisation of volumes of liquid that takes a longer time to reach sterilising conditions
- No activation is required: just expose the unit in a sterilizer and incubate afterward
ProSpore® ampoule is a hermetically sealed, type I borosilicate glass ampoule. The ampoule is filled with a modified soybean casein digest broth containing bromocresol purple acid indicator. Each ampoule also contains a population of G. stearothermophilus spores. Growth is evident by either turbidity and/or a colour change from a purple to or toward yellow. ProSpore® ampoule is a refrigerated item requiring expedited shipping.
Expiration: 18 months from date of manufacture.