Gadolinium ≥99.99%
Catalog # APOSIN1801-1KG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:157.25 g/mol
Boiling Pt:3233 °C
Melting Pt:1312 °C
:GHS02
CAS Number:7440-54-2
Formula:Gd
MDL Number:MFCD00011022