Anti-MALT1 Mouse Monoclonal Antibody [clone: MT1/410] (Alexa Fluor 488)
Catalog # NOVUNBP233054AF488
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen symbol:MALT1
- Clonality:Monoclonal
- Clone:MT1/410
- Conjugation:Alexa Fluor® 488
- ELISA:Yes
- Host:Mouse
- Isotype:IgG kappa
- Reactivity:Human
- Cross adsorption:No
- Form:Purified
- Gene ID:10892
- Storage temperature:Store at 4 °C in the dark
- Tested applications:ELISA
- Size:0,1 ml
- Pk:0,1 mL
Specifications
About this item
MALT1 Monoclonal specifically detects MALT1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: MALT1
Clonality: Monoclonal
Clone: MT1/410
Conjugation: Alexa Fluor 488
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human