Pentyl hexanoate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:540-07-8
:
Formula:C₁₁H₂₂O₂
MDL Number:MFCD00027281
:186.29 g/mol
Merck Index:13,00609
Boiling Pt:225 °C (1013 hPa)
Melting Pt:–47 °C
Density:0.858 g/cm³ (25 °C)
:M-Clarity™ quality level = MQ300
EINECS:208-732-4
Frequently Bought Together