4-Chloro-3-(trifluoromethyl)phenylboronic acid 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:176976-42-4
:
Formula:C₇H₅BClF₃O₂
MDL Number:MFCD03094999
:224.37
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together