5-Bromocytosine 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2240-25-7
:
Formula:C₄H₄BrN₃O
MDL Number:MFCD00056025
:190 g/mol
Melting Pt:240…243 °C
EINECS:218-806-8
:GHS05
:GHS07
Specification Test Results
- Appearance:Light yellow Powder
- Infrared spectrum:Conforms
- Assay:≥98.5 %
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together