Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Ammonium iron (III) citrate, TECHNICAL

Ammonium iron (III) citrate, TECHNICAL

Supplier:   VWR Chemicals
Ammonium iron (III) citrate, TECHNICAL
Supplier:  VWR Chemicals
Certificates
Ammonium iron (III) citrate, TECHNICAL

2 Options Available Below

Product Details & Documents
Synonyms:  Ammonium ferric citrate, Ferric Ammonium Citrate, Ammonium iron (III) citrate
  • CAS Number:1185-57-5
  • Formula:C₆H₅+4y)Fe(x)N(y)O₇
  • MDL Number:MFCD00013099
  • Storage Temperature:Ambient
  • EINECS:214-686-6
Specifications for Product Family

Specification Test Results

  • Assay (calculated as Fe):
    14 - 16 %
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.