Order Entry
United Kingdom
ContactUsLinkComponent
Pesticide reference standard, 2,3,6-Trichlorotoluene 10 µg/ml in methanol, Dr Ehrenstorfer
Pesticide reference standard, 2,3,6-Trichlorotoluene 10 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERLA17787900ME
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, 2,3,6-Trichlorotoluene 10 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERLA17787900ME
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    2077-46-5
  • Description:
    2,3,6-Trichlorotoluene 10 µg/ml in methanol
  • Formula:
    C₇H₅Cl₃
  • Description:
    2,3,6-Trichlorotoluene

Specifications

About this item