5-Isopropyl-2-methylphenol
Catalog # ROTH8808.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pk:10 mL
- Environmentally Preferable:
Specifications
About this item
Synonyms: Carvacrol, 5-Isopropyl-o-cresol, Isothymol, 5-Isopropyl-2-methylphenol
:
:150.22 g/mol
EINECS:207-889-6
UN:3265
CAS Number:499-75-2
Formula:C₁₀H₁₄O
MDL Number:MFCD00002236
Boiling Pt:237
Melting Pt:0.5
Density:0.98 g/cm³ (20 °C)
Flash Pt:>100 °C
ADR:8 III