Human Tagged ORF Clone Lentiviral Particle, BRG1 (SMARCA4)
Catalog # ORIGRC219258L3V
Supplier: OriGene
CAS Number:
Specifications
- Pk:200 µl
- Description:BRG1 (SMARCA4) (NM_003072) human tagged ORF clone lentiviral particle
- Vector details:pLenti-C-Myc-DDK-P2A-Puro (PS100092)
Specifications
About this item
Lenti ORF particles, SMARCA4 (Myc-DDK-tagged)-Human SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 4 (SMARCA4), transcript variant 3, 200 µl, >10^7 TU/ml.
The molecular sequence of this clone aligns with the gene accession number as a point of reference only. However, individual transcript sequences of the same gene can differ through naturally occurring variations (e.g. polymorphisms), each with its own valid existence. This clone is substantially in agreement with the reference, but a complete review of all prevailing variants is recommended prior to use.
This clone was engineered to express the complete ORF with an expression tag. Expression varies depending on the nature of the gene.