Anti-SET1B Rabbit Monoclonal Antibody [clone: E7D1G]
CSTA20055S
- Antibody type:Primary
- Antigen symbol:SET1B
- Clonality:Monoclonal
- Clone:E7D1G
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- ChIP:Yes
- Sequence:CEVDIETEAVAPEERPS(norL)LDEPPLPVG
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Type: Primary
Antigen: SET1B
Clonality: Monoclonal
Clone: E7D1G
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human