Anti-NRF2 Rabbit Monoclonal Antibody [clone: E5F1A]
CSTA20733S
- Antibody type:Primary
- Antigen symbol:NRF2
- Clonality:Monoclonal
- Clone:E5F1A
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- ChIP:Yes
- Sequence:CETGEFLPIQPAQHIQS
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Type: Primary
Antigen: NRF2
Clonality: Monoclonal
Clone: E5F1A
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey