Anti-Androgen Receptor Rabbit Monoclonal Antibody [clone: D6F11] (PE)
Catalog # CSTA8428S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:Androgen Receptor
- Clonality:Monoclonal
- Clone:D6F11
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Sequence:GST-AR(1) 92-194 aa
- Storage temperature:4 °C
- Shipping temperature:Cold
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Androgen Receptor
Clonality: Monoclonal
Clone: D6F11
Conjugation: PE
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human