Anti-INK4A, p16 Rabbit Monoclonal Antibody [clone: D7C1M] (PE)
Catalog # CSTA82548S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:INK4A, p16
- Clonality:Monoclonal
- Clone:D7C1M
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Sequence:NHARIDAAEGPC
- Storage temperature:4 °C
- Shipping temperature:Cold
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: INK4A, p16
Clonality: Monoclonal
Clone: D7C1M
Conjugation: PE
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human