Anti-HIF2-alpha Rabbit Monoclonal Antibody [clone: D6T8V]
Catalog # CSTA59973S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:HIF2-alpha
- Clonality:Monoclonal
- Clone:D6T8V
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- ChIP:Yes
- Sequence:CKTRSAKGFGARGPDVLSPA
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: HIF2-alpha
Clonality: Monoclonal
Clone: D6T8V
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human