Anti-AMOT Rabbit Monoclonal Antibody [clone: D2O4H]
Catalog # CSTA43130S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:AMOT
- Clonality:Monoclonal
- Clone:D2O4H
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Sequence:CATGPGPHRLSIPSLTGNPDKTD
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: AMOT
Clonality: Monoclonal
Clone: D2O4H
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human