Anti-EphB1 Mouse Monoclonal Antibody [clone: 5F10]
CSTA3980S
- Antibody type:Primary
- Antigen symbol:EphB1
- Clonality:Monoclonal
- Clone:5F10
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Sequence:Purified truncated recombinant EphB1-His(aa19-133) is expressed in E.coli strain BL21 (DE3).
- Storage temperature:−20 °C
- Shipping temperature:Cold
- Pk:100 µl
Type: Primary
Antigen: EphB1
Clonality: Monoclonal
Clone: 5F10
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human