Anti-Tuberin/TSC2 (Thr1462) Phosphate Rabbit Monoclonal Antibody [clone: 5B12]
Type: Primary
Antigen: Tuberin/TSC2 (Thr1462) phosphate
Clonality: Monoclonal
Clone: 5B12
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Catalog No:
- CSTA3617T
- CSTA3617S
- Antigen symbol:
- Tuberin/TSC2 (Thr1462) phosphate
- Tuberin/TSC2 (Thr1462) phosphate
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- 5B12
- 5B12
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Sequence:
- CRPRGYtISDSAP
- CRPRGYtISDSAP
- Storage temperature:
- −20 °C
- −20 °C
- Shipping temperature:
- Ambient
- Ambient
Frequently Bought Together