Anti-GRP75 Rabbit Monoclonal Antibody [clone: D13H4]
Supplier: Cell Signaling
Certificates
About this item
Type: Primary
Antigen: GRP75
Clonality: Monoclonal
Clone: D13H4
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Monkey
Specifications
- Catalog No:
- CSTA3593T
- CSTA3593S
- Antigen symbol:
- GRP75
- GRP75
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- D13H4
- D13H4
- Reactivity:
- Monkey, Human, Mouse
- Monkey, Human, Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Sequence:
- CGSGSSGTGEQKEDQKEEKQ
- CGSGSSGTGEQKEDQKEEKQ
- Storage temperature:
- −20 °C
- −20 °C
- Shipping temperature:
- Ambient
- Ambient
Specifications
Frequently Bought Together