Order Entry
United Kingdom
ContactUsLinkComponent
Anti-Cas9 Bacteria Mouse Monoclonal Antibody [clone: 7A9-3A3] (PE)
Anti-Cas9 Bacteria Mouse Monoclonal Antibody [clone: 7A9-3A3] (PE)
Catalog # CSTA35193S
Supplier:  Cell Signaling
Anti-Cas9 Bacteria Mouse Monoclonal Antibody [clone: 7A9-3A3] (PE)
Catalog # CSTA35193S
Supplier:  Cell Signaling

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Cas9 bacteria
  • Clonality:
    Monoclonal
  • Clone:
    7A9-3A3
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Multiple
  • Storage temperature:
    4 °C
  • Shipping temperature:
    Cold
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: Cas9 bacteria
Clonality: Monoclonal
Clone: 7A9-3A3
Conjugation: PE
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Any species