Anti-eIF2-beta Rabbit Polyclonal Antibody
CSTA50537S
- Antibody type:Primary
- Antigen symbol:eIF2-beta
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Sequence:CRKFV(norL)KPPQVVRVGTKKT
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Type: Primary
Antigen: eIF2-beta
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat;Monkey