Anti-IRF-1 Rabbit Monoclonal Antibody [clone: D5E4] (Biotin)
Catalog # CSTA79132S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:IRF-1
- Clonality:Monoclonal
- Clone:D5E4
- Conjugation:Biotin
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Sequence:CDGKGYLLNEPGVQPTSVYGDFS
- Storage temperature:–20 °C
- Shipping temperature:Cold
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: IRF-1
Clonality: Monoclonal
Clone: D5E4
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat