Anti-Rabbit IgG isotype control Rabbit Monoclonal Antibody (Pacific Blue)
Catalog # CSTA9078S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:Rabbit IgG isotype control
- Clonality:Monoclonal
- Conjugation:Pacific Blue™
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Storage temperature:+4 °C
- Shipping temperature:Cold
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Rabbit IgG isotype control
Clonality: Monoclonal
Clone:
Conjugation: Pacific Blue
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: