Anti-liprin beta 1 Rabbit Monoclonal Antibody [clone: D6O4Z]
CSTA67189S
- Antibody type:Primary
- Antigen symbol:liprin beta 1
- Clonality:Monoclonal
- Clone:D6O4Z
- Host:Rabbit
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Sequence:CGTRKVRSSFGRGFFKIKSNKRTAS
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Type: Primary
Antigen: liprin beta 1
Clonality: Monoclonal
Clone: D6O4Z
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Monkey