Anti-EZH2 Rabbit Monoclonal Antibody [clone: D2C9]
Type: Primary
Antigen: EZH2
Clonality: Monoclonal
Clone: D2C9
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Monkey
- Catalog No:
- CSTA5246S
- CSTA5246T
- Antigen symbol:
- EZH2
- EZH2
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- D2C9
- D2C9
- Reactivity:
- Monkey, Human, Rat, Mouse
- Monkey, Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- ChIP:
- Yes
- Yes
- Sequence:
- CKTPPKRPGGRRRGRLPNNSSR
- CKTPPKRPGGRRRGRLPNNSSR
- Storage temperature:
- –20 °C
- –20 °C
- Shipping temperature:
- Cold
- Cold
- Tested applications:
- CUT&RUN
- CUT&RUN
Frequently Bought Together