Anti-RhoC Rabbit Monoclonal Antibody [clone: D40E4]
Type: Primary
Antigen: RhoC
Clonality: Monoclonal
Clone: D40E4
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Monkey
- Catalog No:
- CSTA3430S
- CSTA3430T
- Antigen symbol:
- RhoC
- RhoC
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- D40E4
- D40E4
- Reactivity:
- Monkey, Human, Mouse
- Monkey, Human, Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- Sequence:
- RSEEGRDMANRISAFGYLEC
- RSEEGRDMANRISAFGYLEC
- Storage temperature:
- –20 °C
- –20 °C
- Shipping temperature:
- Ambient
- Ambient
Frequently Bought Together