Anti-CCK4 Rabbit Monoclonal Antibody [clone: D2Z1N]
Catalog # CSTA25618S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:CCK4
- Clonality:Monoclonal
- Clone:D2Z1N
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Sequence:CSPKDRPSFSEIASALGDSTVDSKP
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: CCK4
Clonality: Monoclonal
Clone: D2Z1N
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat