Anti-Phospho-Tyrosine (P-Tyr-1000) MultiMab™ Rabbit Monoclonal Antibody (Magnetic Bead)
Catalog # CSTA14017S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:Phospho-Tyrosine (P-Tyr-1000) MultiMab™
- Clonality:Monoclonal
- Conjugation:Magnetic beads
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Multiple
- Sequence:CXXXKXyXXPXXX
- Storage temperature:+4 °C
- Shipping temperature:Cold
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: Phospho-Tyrosine (P-Tyr-1000) MultiMab™
Clonality: Monoclonal
Clone:
Conjugation: Magnetic Bead
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Any species