Anti-syntenin Rabbit Monoclonal Antibody [clone: E2I9L]
Catalog # CSTA27964S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:syntenin
- Clonality:Monoclonal
- Clone:E2I9L
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Sequence:O00560 L7-L117
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: syntenin
Clonality: Monoclonal
Clone: E2I9L
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human