Anti-NOP2 Rabbit Monoclonal Antibody [clone: E9Z4D]
Catalog # CSTA90709S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:NOP2
- Clonality:Monoclonal
- Clone:E9Z4D
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Sequence:CEPFVLPPAGE(norL)EQDAQAPD
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: NOP2
Clonality: Monoclonal
Clone: E9Z4D
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey