Anti-Rabbit IgG isotype control Rabbit Monoclonal Antibody
Catalog # CSTA66362S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:Rabbit IgG isotype control
- Clonality:Monoclonal
- Host:Rabbit
- Isotype:IgG
- Storage temperature:–20 °C
- Shipping temperature:Cold
- Tested applications:CUT&RUN
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Rabbit IgG isotype control
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: