Anti-NR2C2 Rabbit Monoclonal Antibody [clone: E1T8N]
Catalog # CSTA31646S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:NR2C2
- Clonality:Monoclonal
- Clone:E1T8N
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Sequence:TSPSPRIQIISTDSAVAC
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: NR2C2
Clonality: Monoclonal
Clone: E1T8N
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey