Anti-ATF7 (Thr53) phosphate Rabbit Monoclonal Antibody [clone: A8J7P]
Catalog # CSTA15411S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:ATF7 (Thr53) phosphate
- Clonality:Monoclonal
- Clone:A8J7P
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Antigen synonyms:ATF-2 (Thr71) phosphate
- Sequence:DQTPtPTRFLKC
- Storage temperature:–20 °C
- Shipping temperature:Cold
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: ATF7 (Thr53) phosphate
Clonality: Monoclonal
Clone: A8J7P
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey