Anti-LysRS Rabbit Monoclonal Antibody [clone: D16D1]
CSTA9990S
- Antibody type:Primary
- Antigen symbol:LysRS
- Clonality:Monoclonal
- Clone:D16D1
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Sequence:CTDSNNIKEVLLFPAMKPEDKKE
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Type: Primary
Antigen: LysRS
Clonality: Monoclonal
Clone: D16D1
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey