Anti-p70 S6k1 (Thr398) phosphate drosophila Rabbit Polyclonal Antibody
Catalog # CSTA9209S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:p70 S6k1 (Thr398) phosphate drosophila
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Drosophila melanogaster,Human
- Western blot:Yes
- Sequence:NLIFQGFtYVAPSIC
- Storage temperature:–20 °C
- Shipping temperature:Cold
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: p70 S6k1 (Thr398) phosphate drosophila
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, D. melanogaster