Anti-ULK1 Rabbit Monoclonal Antibody [clone: D8H5]
Supplier: Cell Signaling
Certificates
About this item
Type: Primary
Antigen: ULK1
Clonality: Monoclonal
Clone: D8H5
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey
Specifications
- Catalog No:
- CSTA8054S
- CSTA8054T
- Antigen symbol:
- ULK1
- ULK1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- D8H5
- D8H5
- Reactivity:
- Monkey, Human, Rat, Mouse
- Monkey, Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Sequence:
- CRNLRGSPKLPDFLQRNPL
- CRNLRGSPKLPDFLQRNPL
- Storage temperature:
- –20 °C
- –20 °C
- Shipping temperature:
- Cold
- Cold
Specifications
Frequently Bought Together