Anti-GRK6 Rabbit Monoclonal Antibody [clone: D1A4]
CSTA5878S
- Antibody type:Primary
- Antigen symbol:GRK6
- Clonality:Monoclonal
- Clone:D1A4
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Sequence:CVAFLDGVAEYEVTPDDKRKA
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Type: Primary
Antigen: GRK6
Clonality: Monoclonal
Clone: D1A4
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat